(4-(4-(Bis(4-fluorophenyl)methyl)piperazin-1-yl)phenyl)methanol ≥95%

Supplier: Matrix Scientific

071881-1G 071881-5G 071881-10G
102521-878EA 213.85 USD
102521-878 102521-880 102521-882
(4-(4-(Bis(4-fluorophenyl)methyl)piperazin-1-yl)phenyl)methanol ≥95%
(4-(4-(Bis(4-fluorophenyl)methyl)piperazin-1-yl)phenyl)methanol

Hazard: Irritant

Formula: C₂₄H₂₄F₂N₂O
CAS Number: 914349-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR