2'-(Trifluoromethyl)biphenyl-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 2'-(Trifluoromethyl)-[1,1'-biphenyl]-3-carbaldehyde

071879-1G 071879-5G 071879-10G
102521-866EA 355.38 USD
102521-866 102521-868 102521-870
2'-(Trifluoromethyl)biphenyl-3-carbaldehyde ≥95%
2'-(Trifluoromethyl)biphenyl-3-carbaldehyde

Hazard: Irritant

Formula: C₁₄H₉F₃O
MW: 250.2159 g/mol
MDL Number: MFCD03424642
CAS Number: 675596-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR