4'-(Trifluoromethyl)biphenyl-3-carboxylic acid ≥95%
Supplier: Matrix Scientific
071878-1G
071878-5G
071878-10G
102521-860EA
355.38
USD
102521-860
102521-862
102521-864
4'-(Trifluoromethyl)biphenyl-3-carboxylic acid ≥95%
4'-(Trifluoromethyl)biphenyl-3-carboxylic acid
Hazard: Irritant
Formula:
C₁₄H₉F₃O₂ MW: 266.22 g/mol |
MDL Number:
MFCD03424604 CAS Number: 199528-28-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...