2'-(Trifluoromethyl)biphenyl-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2'-(Trifluoromethyl)-[1,1'-biphenyl]-4-carboxylic acid

071876-1G 071876-5G 071876-10G
102521-848EA 355.38 USD
102521-848 102521-850 102521-852
2'-(Trifluoromethyl)biphenyl-4-carboxylic acid ≥95%
2'-(Trifluoromethyl)biphenyl-4-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₉F₃O₂
MW: 266.22 g/mol
MDL Number: MFCD03424605
CAS Number: 198205-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR