2-Naphthalen-1-ylpiperazine ≥95%

Supplier: Matrix Scientific

071875-1G 071875-5G
102521-844EA 640 USD
102521-844 102521-846
2-Naphthalen-1-ylpiperazine ≥95%
2-Naphthalen-1-ylpiperazine

Hazard: Irritant

Formula: C₁₄H₁₆N₂
CAS Number: 910444-80-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR