3-Phenylthiomorpholine ≥95%

Supplier: Matrix Scientific

071872-1G 071872-5G
102521-830EA 498.46 USD
102521-830 102521-832
3-Phenylthiomorpholine ≥95%
3-Phenylthiomorpholine

Hazard: Irritant

Formula: C10H13NS
CAS Number: 141849-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR