4-(4-(Bis(4-fluorophenyl)methyl)piperazin-1-yl)benzaldehyde ≥95%

Supplier: Matrix Scientific

071868-1G 071868-5G 071868-10G
102521-806EA 213.85 USD
102521-806 102521-808 102521-810
4-(4-(Bis(4-fluorophenyl)methyl)piperazin-1-yl)benzaldehyde ≥95%
4-(4-(Bis(4-fluorophenyl)methyl)piperazin-1-yl)benzaldehyde

Hazard: Irritant

Formula: C₂₄H₂₂F₂N₂O
CAS Number: 914349-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR