4-(Boc-amino)-3-iodopyridine ≥95%

Supplier: Matrix Scientific

071866-1G 071866-5G 071866-10G
102521-796EA 123.08 USD
102521-796 102521-798 102521-800
4-(Boc-amino)-3-iodopyridine ≥95%
4-(Boc-amino)-3-iodopyridine

Hazard: Irritant

Formula: C₁₀H₁₃IN₂O₂
MDL Number: MFCD04973409
CAS Number: 211029-67-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR