3-Bromo-4-pyridone ≥95%
Supplier: Matrix Scientific
071865-1G
071865-5G
102521-790EA
53.85
USD
102521-790
102521-792
3-Bromo-4-pyridone ≥95%
3-Bromo-4-pyridone
Hazard: Irritant
Formula:
C₅H₄BrNO |
MDL Number:
MFCD01646107 CAS Number: 36953-41-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...