3-(3-Bromo-4-methoxyphenyl)pyridine ≥95%

Supplier: Matrix Scientific

071863-1G 071863-5G 071863-10G
102521-778EA 355.38 USD
102521-778 102521-780 102521-782
3-(3-Bromo-4-methoxyphenyl)pyridine ≥95%
3-(3-Bromo-4-methoxyphenyl)pyridine

Hazard: Irritant

Formula: C₁₂H₁₀BrNO
MW: 264.1200 g/mol
MDL Number: MFCD05864836
CAS Number: 914349-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR