2-(4-Ethylpiperazin-1-ylmethyl)benzoic acid ≥95%

Supplier: Matrix Scientific

071862-1G 071862-5G 071862-10G
102521-772EA 533.85 USD
102521-772 102521-774 102521-776
2-(4-Ethylpiperazin-1-ylmethyl)benzoic acid ≥95%
2-(4-Ethylpiperazin-1-ylmethyl)benzoic acid

Hazard: Irritant

Formula: C₁₄H₂₀N₂O₂
CAS Number: 914349-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR