2-(4-Methylpiperazin-1-ylmethyl)benzoic acid ≥95%

Supplier: Matrix Scientific

071861-1G 071861-5G 071861-10G
102521-766EA 533.85 USD
102521-766 102521-768 102521-770
2-(4-Methylpiperazin-1-ylmethyl)benzoic acid ≥95%
2-(4-Methylpiperazin-1-ylmethyl)benzoic acid

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₂
MW: 234.2900 g/mol
MDL Number: MFCD06797777
CAS Number: 514209-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR