3-(Morpholinomethyl)benzoic acid ≥95%

Supplier: Matrix Scientific

071860-1G 071860-5G 071860-10G
102521-760EA 533.85 USD
102521-760 102521-762 102521-764
3-(Morpholinomethyl)benzoic acid ≥95%
3-(Morpholinomethyl)benzoic acid

Hazard: Irritant

Formula: C₁₂H₁₅NO₃
MW: 221.2524 g/mol
MDL Number: MFCD05841588
CAS Number: 67451-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR