3-(4-Methylpiperazin-1-ylmethyl)benzoic acid ≥95%

Supplier: Matrix Scientific

071859-1G 071859-5G 071859-10G
102521-754EA 463.08 USD
102521-754 102521-756 102521-758
3-(4-Methylpiperazin-1-ylmethyl)benzoic acid ≥95%
3-(4-Methylpiperazin-1-ylmethyl)benzoic acid

Hazard: Irritant

Formula: C13H18N2O2
CAS Number: 514209-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR