2-((4-(tert-Butoxycarbonyl)piperazin-1-yl)methyl)benzoic acid ≥95%

Supplier: Matrix Scientific

071858-1G 071858-5G 071858-10G
102521-748EA 533.85 USD
102521-748 102521-750 102521-752
2-((4-(tert-Butoxycarbonyl)piperazin-1-yl)methyl)benzoic acid ≥95%
2-((4-(tert-Butoxycarbonyl)piperazin-1-yl)methyl)benzoic acid

Hazard: Irritant

Formula: C₁₇H₂₄N₂O₄
CAS Number: 914349-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR