4-(4-(tert-Butoxycarbonyl)piperazin-1-yl)benzoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: Boc-piperazine-benzoic acid

071857-1G 071857-5G 071857-10G
102521-742EA 89.23 USD
102521-742 102521-744 102521-746
4-(4-(tert-Butoxycarbonyl)piperazin-1-yl)benzoic acid ≥95%
4-(4-(tert-Butoxycarbonyl)piperazin-1-yl)benzoic acid

Hazard: Irritant

Formula: C₁₆H₂₂N₂O₄
MW: 306.3569 g/mol
MDL Number: MFCD04115067
CAS Number: 162046-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR