3-(4-Pyridyl)benzaldehyde ≥95%
Supplier: Matrix Scientific
Synonyms:
3-(Pyridin-4-yl)benzaldehyde, 3-Pyrid-4-ylbenzaldehyde
071855-1G
071855-5G
071855-10G
102521-730EA
284.62
USD
102521-730
102521-732
102521-734
3-(4-Pyridyl)benzaldehyde ≥95%
3-(4-Pyridyl)benzaldehyde
Hazard: Irritant
Formula:
C₁₂H₉NO MW: 183.2060 g/mol |
MDL Number:
MFCD02684105 CAS Number: 208190-04-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...