2-(2-Aminopyrimidin-5-yl)benzaldehyde ≥95%

Supplier: Matrix Scientific

071854-1G 071854-5G 071854-10G
102521-724EA 355.38 USD
102521-724 102521-726 102521-728
2-(2-Aminopyrimidin-5-yl)benzaldehyde ≥95%
2-(2-Aminopyrimidin-5-yl)benzaldehyde

Hazard: Irritant

Formula: C₁₁H₉N₃O
CAS Number: 914349-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR