4-(Pyrimidin-5-yl)benzaldehyde ≥95%

Supplier: Matrix Scientific

071853-1G 071853-5G 071853-10G
102521-718EA 249.23 USD
102521-718 102521-720 102521-722
4-(Pyrimidin-5-yl)benzaldehyde ≥95%
4-(Pyrimidin-5-yl)benzaldehyde

Hazard: Irritant

Formula: C₁₁H₈N₂O
MW: 184.1940 g/mol
MDL Number: MFCD02684108
CAS Number: 198084-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR