2-(3-Pyridyl)benzaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(Pyridin-3-yl)benzaldehyde

071850-1G 071850-5G 071850-10G
102521-700EA 320 USD
102521-700 102521-702 102521-704
2-(3-Pyridyl)benzaldehyde ≥95%
2-(3-Pyridyl)benzaldehyde

Hazard: Irritant

Formula: C₁₂H₉NO
MDL Number: MFCD02684097
CAS Number: 176690-44-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR