4-(6-Bromopyridin-2-yl)benzaldehyde ≥95%

Supplier: Matrix Scientific

071849-1G 071849-5G 071849-10G
102521-694EA 533.85 USD
102521-694 102521-696 102521-698
4-(6-Bromopyridin-2-yl)benzaldehyde ≥95%
4-(6-Bromopyridin-2-yl)benzaldehyde

Hazard: Irritant

Formula: C₁₂H₈BrNO
MDL Number: MFCD05864828
CAS Number: 588727-65-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR