4-(4-Pyridyl)benzaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(Pyridin-4-yl)benzaldehyde

071848-1G 071848-5G 071848-10G
102521-688EA 249.23 USD
102521-688 102521-690 102521-692
4-(4-Pyridyl)benzaldehyde ≥95%
4-(4-Pyridyl)benzaldehyde

Hazard: Irritant

Formula: C₁₂H₉NO
MW: 183.21 g/mol
MDL Number: MFCD02684110
CAS Number: 99163-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR