(4-(4-Ethylpiperazin-1-yl)phenyl)methanol ≥95%

Supplier: Matrix Scientific

071846-1G 071846-5G 071846-10G
102521-676EA 284.62 USD
102521-676 102521-678 102521-680
(4-(4-Ethylpiperazin-1-yl)phenyl)methanol ≥95%
(4-(4-Ethylpiperazin-1-yl)phenyl)methanol

Hazard: Irritant

Formula: C₁₃H₂₀N₂O
CAS Number: 402745-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR