(2-Imidazol-1-yl-phenyl)methanol ≥95%

Supplier: Matrix Scientific

071844-1G 071844-5G 071844-10G
102521-664EA 284.62 USD
102521-664 102521-666 102521-668
(2-Imidazol-1-yl-phenyl)methanol ≥95%
(2-Imidazol-1-yl-phenyl)methanol

Hazard: Irritant

Formula: C₁₀H₁₀N₂O
MW: 174.2000 g/mol
MDL Number: MFCD05864823
CAS Number: 25373-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR