[2-(4-Benzylpiperazin-1-yl)phenyl]methanol ≥95%
Supplier: Matrix Scientific
071843-1G
071843-5G
071843-10G
102521-658EA
284.62
USD
102521-658
102521-660
102521-662
[2-(4-Benzylpiperazin-1-yl)phenyl]methanol ≥95%
[2-(4-Benzylpiperazin-1-yl)phenyl]methanol
Hazard: Irritant
Formula:
C₁₈H₂₂N₂O MW: 282.3801 g/mol |
MDL Number:
MFCD01764934 CAS Number: 261178-24-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...