(4-Morpholin-4-yl-phenyl)methanol ≥95%

Supplier: Matrix Scientific

071840-1G 071840-5G
102521-642EA 143.08 USD
102521-642 102521-644
(4-Morpholin-4-yl-phenyl)methanol ≥95%
(4-Morpholin-4-yl-phenyl)methanol

Hazard: Irritant

Formula: C₁₁H₁₅NO₂
MW: 193.2423 g/mol
MDL Number: MFCD01057413
CAS Number: 280556-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR