(4-(2-Methyl-1H-imidazol-1-yl)phenyl)methanol ≥95%

Supplier: Matrix Scientific

071839-1G 071839-5G 071839-10G
102521-636EA 284.62 USD
102521-636 102521-638 102521-640
(4-(2-Methyl-1H-imidazol-1-yl)phenyl)methanol ≥95%
(4-(2-Methyl-1H-imidazol-1-yl)phenyl)methanol

Hazard: Irritant

Formula: C₁₁H₁₂N₂O
MW: 188.23 g/mol
MDL Number: MFCD05864819
CAS Number: 167758-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR