(2-(2-Methyl-1H-imidazol-1-yl)phenyl)methanol ≥95%

Supplier: Matrix Scientific

071838-1G 071838-5G 071838-10G
102521-630EA 284.62 USD
102521-630 102521-632 102521-634
(2-(2-Methyl-1H-imidazol-1-yl)phenyl)methanol ≥95%
(2-(2-Methyl-1H-imidazol-1-yl)phenyl)methanol

Hazard: Irritant

Formula: C₁₁H₁₂N₂O
MDL Number: MFCD05864818
CAS Number: 914349-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR