3-(5-Nitropyridin-2-yl)benzoic acid ≥95%

Supplier: Matrix Scientific

071834-1G 071834-5G 071834-10G
102521-606EA 355.38 USD
102521-606 102521-608 102521-610
3-(5-Nitropyridin-2-yl)benzoic acid ≥95%
3-(5-Nitropyridin-2-yl)benzoic acid

Hazard: Irritant

Formula: C12H8N2O4
CAS Number: 864075-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR