4-Pyrimidin-2-ylbenzoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Pyrimidin-2-yl-benzoic acid

071831-1G 071831-5G 071831-10G
102521-588EA 400 USD
102521-588 102521-590 102521-592
4-Pyrimidin-2-ylbenzoic acid ≥95%
4-Pyrimidin-2-ylbenzoic acid

Hazard: Irritant

Formula: C₁₁H₈N₂O₂
MW: 200.1900 g/mol
MDL Number: MFCD01318668
CAS Number: 199678-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR