3-Pyrimidin-2-ylbenzoic acid ≥95%

Supplier: Matrix Scientific

071830-1G 071830-5G 071830-10G
102521-582EA 241.54 USD
102521-582 102521-584 102521-586
3-Pyrimidin-2-ylbenzoic acid ≥95%
3-Pyrimidin-2-ylbenzoic acid

Hazard: Irritant

Formula: C₁₁H₈N₂O₂
CAS Number: 579476-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR