3-(6-Bromopyridin-2-yl)benzoic acid ≥95%

Supplier: Matrix Scientific

071825-1G 071825-5G 071825-10G
102521-552EA 533.85 USD
102521-552 102521-554 102521-556
3-(6-Bromopyridin-2-yl)benzoic acid ≥95%
3-(6-Bromopyridin-2-yl)benzoic acid

Hazard: Irritant

Formula: C12H8BrNO2
CAS Number: 914349-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR