3-Pyrid-4-ylbenzoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(Pyridin-4-yl)benzoic acid

071824-1G 071824-5G 071824-10G
102521-546EA 320 USD
102521-546 102521-548 102521-550
3-Pyrid-4-ylbenzoic acid ≥95%
3-Pyrid-4-ylbenzoic acid

Hazard: Irritant

Formula: C₁₂H₉NO₂
MW: 199.2054 g/mol
MDL Number: MFCD05864808
CAS Number: 4385-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR