3-Pyrid-2-ylbenzoic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
3-(Pyridin-2-yl)benzoic acid
071823-1G
071823-5G
071823-10G
102521-540EA
153.85
USD
102521-540
102521-542
102521-544
3-Pyrid-2-ylbenzoic acid ≥95%
3-Pyrid-2-ylbenzoic acid
Hazard: Irritant
Formula:
C₁₂H₉NO₂ MW: 199.2054 g/mol |
MDL Number:
MFCD05864807 CAS Number: 4467-07-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...