Ethyl-3-bromoimidazo[1,2-a]pyridine-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-bromoimidazo[1,2-a]pyridine-2-carboxylate

071821-1G 071821-5G
102521-528EA 150.77 USD
102521-528 102521-530
Ethyl-3-bromoimidazo[1,2-a]pyridine-2-carboxylate ≥95%
Ethyl 3-bromoimidazo[1,2-a]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₉BrN₂O₂
MW: 269.1000 g/mol
MDL Number: MFCD05864806
CAS Number: 143982-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR