Ethyl imidazo[2,1-b]thiazole-6-carboxylate ≥95%

Supplier: Matrix Scientific

071819-1G 071819-5G 071819-10G
102521-516EA 355.38 USD
102521-516 102521-518 102521-520
Ethyl imidazo[2,1-b]thiazole-6-carboxylate ≥95%
Ethyl imidazo[2,1-b]thiazole-6-carboxylate

Hazard: Irritant

Formula: C₈H₈N₂O₂S
CAS Number: 64951-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR