Ethyl-6-aminoimidazo[1,2-a]pyridine-2-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 6-aminoimidazo[1,2-a]pyridine-2-carboxylate
071818-1G
071818-5G
071818-10G
102521-510EA
218.46
USD
102521-510
102521-512
102521-514
Ethyl-6-aminoimidazo[1,2-a]pyridine-2-carboxylate ≥95%
Ethyl 6-aminoimidazo[1,2-a]pyridine-2-carboxylate
Hazard: Irritant
Formula:
C₁₀H₁₁N₃O₂ MW: 205.2132 g/mol |
MDL Number:
MFCD05864804 CAS Number: 158980-21-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...