5-(2,5-Dimethoxyphenyl)-1H-pyrazole-3-carboxylic acid ≥95%
Supplier: Matrix Scientific
071817-1G
071817-5G
071817-10G
102521-504EA
533.85
USD
102521-504
102521-506
102521-508
5-(2,5-Dimethoxyphenyl)-1H-pyrazole-3-carboxylic acid ≥95%
5-(2,5-Dimethoxyphenyl)-1H-pyrazole-3-carboxylic acid
Hazard: Irritant
Formula:
C₁₂H₁₂N₂O₄ MW: 248.2347 g/mol |
MDL Number:
MFCD05170041 CAS Number: 882238-14-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...