5-(4-(Benzyloxy)phenyl)-1H-pyrazole-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

071815-1G 071815-5G 071815-10G
102521-492EA 533.85 USD
102521-492 102521-494 102521-496
5-(4-(Benzyloxy)phenyl)-1H-pyrazole-3-carboxylic acid ≥95%
5-(4-(Benzyloxy)phenyl)-1H-pyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₇H₁₄N₂O₃
CAS Number: 795260-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR