5-(2,4-Dichloro-5-fluorophenyl)-1H-pyrazole-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

071814-1G 071814-5G 071814-10G
102521-486EA 533.85 USD
102521-486 102521-488 102521-490
5-(2,4-Dichloro-5-fluorophenyl)-1H-pyrazole-3-carboxylic acid ≥95%
5-(2,4-Dichloro-5-fluorophenyl)-1H-pyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₅Cl₂FN₂O₂
CAS Number: 1119299-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR