5-(Naphthalen-2-yl)-1H-pyrazole-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

071813-1G 071813-5G 071813-10G
102521-480EA 533.85 USD
102521-480 102521-482 102521-484
5-(Naphthalen-2-yl)-1H-pyrazole-3-carboxylic acid ≥95%
5-(Naphthalen-2-yl)-1H-pyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₀N₂O₂
CAS Number: 164295-94-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR