5-Thiophen-2-yl-1H-pyrazole-3-carboxylic acid ≥95%
Supplier: Matrix Scientific
071812-1G
071812-5G
071812-10G
102521-474EA
533.85
USD
102521-474
102521-476
102521-478
5-Thiophen-2-yl-1H-pyrazole-3-carboxylic acid ≥95%
5-Thiophen-2-yl-1H-pyrazole-3-carboxylic acid
Hazard: Irritant
Formula:
C8H6N2O2S MW: 194.2104 g/mol |
MDL Number:
MFCD03030176 CAS Number: 182415-24-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...