5-(3-Acetamidophenyl)-1H-pyrazole-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

071810-1G 071810-5G 071810-10G
102521-462EA 533.85 USD
102521-462 102521-464 102521-466
5-(3-Acetamidophenyl)-1H-pyrazole-3-carboxylic acid ≥95%
5-(3-Acetamidophenyl)-1H-pyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₁N₃O₃
CAS Number: 1240061-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR