5-(6-Methoxynaphthalen-2-yl)-1H-pyrazole-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

071809-1G 071809-5G 071809-10G
102521-456EA 533.85 USD
102521-456 102521-458 102521-460
5-(6-Methoxynaphthalen-2-yl)-1H-pyrazole-3-carboxylic acid ≥95%
5-(6-Methoxynaphthalen-2-yl)-1H-pyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₂N₂O₃
MW: 268.2674 g/mol
MDL Number: MFCD05864800
CAS Number: 1257877-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR