5-(4-Chlorophenyl)-1H-pyrazole-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

071808-1G 071808-5G 071808-10G
102521-450EA 533.85 USD
102521-450 102521-452 102521-454
5-(4-Chlorophenyl)-1H-pyrazole-3-carboxylic acid ≥95%
5-(4-Chlorophenyl)-1H-pyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₇ClN₂O₂
MW: 222.6300 g/mol
MDL Number: MFCD03715908
CAS Number: 54006-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR