5-(4-Nitrophenyl)-1H-pyrazole-3-carboxylic acid ≥95%
Supplier: Matrix Scientific
071807-1G
071807-5G
071807-10G
102521-444EA
252.31
USD
102521-444
102521-446
102521-448
5-(4-Nitrophenyl)-1H-pyrazole-3-carboxylic acid ≥95%
5-(4-Nitrophenyl)-1H-pyrazole-3-carboxylic acid
Hazard: Irritant
Formula:
C₁₀H₇N₃O₄ MW: 233.1803 g/mol |
MDL Number:
MFCD03086935 CAS Number: 189083-63-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...