5-(3-Chlorophenyl)-1H-pyrazole-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

071804-1G 071804-5G 071804-10G
102521-426EA 533.85 USD
102521-426 102521-428 102521-430
5-(3-Chlorophenyl)-1H-pyrazole-3-carboxylic acid ≥95%
5-(3-Chlorophenyl)-1H-pyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₇ClN₂O₂
MW: 222.6300 g/mol
MDL Number: MFCD05170019
CAS Number: 595610-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR