a-Tosyl(4-methoxybenzyl)isocyanide ≥95%

Supplier: Matrix Scientific

071802-1G 071802-5G 071802-10G
102521-416EA 533.85 USD
102521-416 102521-418 102521-420
a-Tosyl(4-methoxybenzyl)isocyanide ≥95%
a-Tosyl(4-methoxybenzyl)isocyanide

Hazard: Irritant

Formula: C₁₆H₁₅NO₃S
MDL Number: MFCD04114785
CAS Number: 263389-54-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR