a-Tosyl(3-methoxybenzyl)isocyanide ≥95%

Supplier: Matrix Scientific

071801-1G 071801-5G
102521-412EA 498.46 USD
102521-412 102521-414
a-Tosyl(3-methoxybenzyl)isocyanide ≥95%
a-Tosyl(3-methoxybenzyl)isocyanide

Hazard: Irritant

Formula: C₁₆H₁₅NO₃S
MDL Number: MFCD04114786
CAS Number: 394655-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR