a-Tosyl(4-chlorobenzyl)isocyanide ≥95%
Supplier: Matrix Scientific
071800-1G
071800-5G
071800-10G
102521-406EA
533.85
USD
102521-406
102521-408
102521-410
a-Tosyl(4-chlorobenzyl)isocyanide ≥95%
a-Tosyl(4-chlorobenzyl)isocyanide
Hazard: Irritant
Formula:
C₁₅H₁₂ClNO₂S |
CAS Number:
918892-30-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...